Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270880.1 | NP_001257809.1 | 6 | 1227 | ||
NM_001270881.1 | NP_001257810.1 | 6 | 1170 | |||
NM_138335.2 | NP_612208.1 | 7 | 1329 | |||
NR_073094.1 | 7 | 1234 | ||||
NR_073095.1 | 4 | 885 | ||||
GenBank mRNA | AF247786.1 | - | 7 | 1290 | ||
AK055639.1 | - | 7 | 1249 | |||
AL834506.1 | - | 4 | 778 | |||
AY173948.1 | - | 6 | 1164 | |||
BC015532.2 | - | 6 | 1157 | 3914747 |