Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277.2 | NP_001268.2 | 1 | 546 | ||
NM_212469.1 | NP_997634.1 | 1 | 546 | |||
XM_017017147.1 | XP_016872636.1 | 1 | 544 | |||
GenBank mRNA | AI809679.1 | - | 2 | 200 | ||
BC036471.2 | - | 1 | 424 | 5265994 | ||
BG703955.1 | - | 552 | ||||
BG723115.1 | - | 1 | 600 | |||
D10704.1 | - | 1 | 356 |