Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146726.1 | NP_001140198.1 | 5, 6 | 870 | ||
NM_138379.2 | NP_612388.2 | 6, 7 | 954 | |||
XM_017010021.1 | XP_016865510.1 | 4, 5 | 760 | |||
GenBank mRNA | AK131025.1 | - | 2, 3 | 212 | ||
AK301337.1 | - | 5, 6 | 841 | |||
AK312746.1 | - | 6, 7 | 924 | |||
BC008988.1 | - | 6, 7 | 954 | 4184237 | ||
JX049980.1 | - | 6, 7 | 897 | |||
KU178832.1 | - | 6, 7 | 897 | |||
KU178833.1 | - | 5, 6 | 816 |