Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207019.2 | NP_001193948.2 | 7 | 481 | ||
NM_001220500.1 | NP_001207429.1 | 8 | 617 | |||
NM_002002.4 | NP_001993.2 | 8 | 613 | |||
XM_005272462.4 | XP_005272519.1 | 8 | 1218 | |||
GenBank mRNA | AK223207.1 | - | 8 | 582 | ||
AK310071.1 | - | 7 | 762 | |||
AK310707.1 | - | 8 | 583 | |||
BC014108.2 | - | 8 | 578 | 4309266 | ||
BC062591.1 | - | 8 | 571 | 5216918 | ||
BC064417.1 | - | 7 | 895 | |||
M14766.1 | - | 8 | 585 | |||
M15059.1 | - | 8 | 613 | |||
X04772.1 | - | 8 | 613 |