Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318049.1 | NP_001304978.1 | 8 | 2658 | ||
NM_001318050.1 | NP_001304979.1 | 6 | 2379 | |||
NM_138969.3 | NP_620419.2 | 7 | 2511 | |||
XM_011517479.1 | XP_011515781.1 | 7 | 2526 | |||
GenBank mRNA | AK057667.1 | - | 7 | 1854 | ||
AY444559.1 | - | 6 | 1778 | |||
BC037219.1 | - | 7 | 1858 | 5181472 | ||
BC064525.1 | - | 7 | 1830 | 5752917 |