Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159770.1 | NP_001153242.1 | 10 | 1712 | ||
NM_139177.3 | NP_631916.2 | 10 | 1691 | |||
XM_005257134.3 | XP_005257191.1 | 10 | 1778 | |||
XM_006721754.3 | XP_006721817.1 | 10 | 1799 | |||
XM_006721755.2 | XP_006721818.1 | 11 | 2003 | |||
XM_006721757.2 | XP_006721820.1 | 9 | 1541 | |||
XM_011524493.1 | XP_011522795.1 | 11 | 2094 | |||
XM_011524494.2 | XP_011522796.1 | 10 | 1745 | |||
XM_017024330.1 | XP_016879819.1 | 11 | 1982 | |||
XM_017024331.1 | XP_016879820.1 | 11 | 2073 | |||
XM_017024332.1 | XP_016879821.1 | 10 | 1688 | |||
XM_017024333.1 | XP_016879822.1 | 9 | 1652 | |||
XM_017024334.1 | XP_016879823.1 | 9 | 1628 | |||
XM_017024335.1 | XP_016879824.1 | 9 | 1520 | |||
XM_017024336.1 | XP_016879825.1 | 11 | 1892 | |||
XM_017024337.1 | XP_016879826.1 | 11 | 1805 | |||
GenBank mRNA | AF331643.1 | - | 10 | 1653 | ||
AK095227.1 | - | 10 | 1686 | |||
BC035631.1 | - | 10 | 1670 | 5219031 |