Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145288.1 | NP_001138760.1 | 9, 10 | 1468 | ||
NM_139319.2 | NP_647480.1 | 10, 11 | 1618 | |||
GenBank mRNA | AJ459241.1 | - | 10, 11 | 1481 | ||
AK128319.1 | - | 10, 11 | 1618 | |||
BC117229.1 | - | 9, 10 | 1179 | 40125780 | ||
BC143396.1 | - | 9, 10 | 1179 | 9051903 |