Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KGFLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003674.2 | 3 | 1890 | |||
NR_047527.1 | 4 | 2000 | ||||
GenBank mRNA | AY098593.1 | - | 4 | 1821 | ||
CN274734.1 | - | 1 | 320 |
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002009.3 | NP_002000.1 | 4 | 2144 | ||
GenBank mRNA | M60828.1 | - | 4 | 2062 |
Gene Symbol: KGFLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003670.1 | 3 | 1988 | |||
GenBank mRNA | AY098594.1 | - | 3 | 1880 |