Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256385.1 | NP_001243314.1 | 9 | 1170 | ||
NM_001256386.1 | NP_001243315.1 | 9 | 1170 | |||
NM_001256387.1 | NP_001243316.1 | 8 | 1113 | |||
NM_144620.3 | NP_653221.1 | 9 | 1153 | |||
GenBank mRNA | AK096892.1 | - | 9 | 1096 | ||
AL833417.1 | - | 9 | 1118 | |||
AY358131.1 | - | 8 | 1086 | |||
BC009613.1 | - | 9 | 1095 | 4245944 |