Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006723039.3 | XP_006723102.1 | 4 | 2438 | ||
XM_011526503.2 | XP_011524805.1 | 5 | 2498 | |||
XM_011526504.2 | XP_011524806.1 | 4 | 2312 | |||
XM_011526505.2 | XP_011524807.1 | 6 | 2470 | |||
XM_011526507.2 | XP_011524809.1 | 3 | 2146 | |||
XM_011526508.2 | XP_011524810.1 | 2 | 2104 | |||
XM_011526510.2 | XP_011524812.1 | 3 | 2283 | |||
XM_017026337.1 | XP_016881826.1 | 5 | 2403 | |||
XM_017026338.1 | XP_016881827.1 | 2 | 2058 | |||
GenBank mRNA | AK056695.1 | - | 1 | 1725 | ||
BC026210.2 | - | 5 | 2322 | 3028503 |