Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282692.1 | NP_001269621.1 | 6, 7 | 1356 | ||
NM_001282693.1 | NP_001269622.1 | 7, 8 | 1350 | |||
NM_001282694.1 | NP_001269623.1 | 6, 7 | 1161 | |||
NM_002021.2 | NP_002012.1 | 7, 8 | 1341 | |||
XM_005245037.3 | XP_005245094.1 | 5, 6 | 970 | |||
XM_006711241.3 | XP_006711304.1 | 8, 9 | 1245 | |||
XM_006711242.3 | XP_006711305.1 | 7, 8 | 1350 | |||
GenBank mRNA | AK315100.1 | - | 7, 8 | 1300 | ||
BC047129.1 | - | 6, 7 | 1322 | 5186727 | ||
M64082.1 | - | 7, 8 | 1289 |