Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145176.2 | NP_660159.1 | 3 | 1668 | ||
GenBank mRNA | AK056554.1 | - | 2 | 407 | ||
AK122628.1 | - | 3 | 1660 | |||
AK226098.1 | - | 3 | 1539 | |||
AL833602.1 | - | 3 | 1652 | |||
AY046419.1 | - | 3 | 1630 | |||
BC070149.1 | - | 3 | 1602 | 30371087 | ||
BX648889.1 | - | 4 | 803 |