Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168322.1 | NP_001161794.1 | 4 | 658 | ||
NM_001168323.1 | NP_001161795.1 | 3 | 556 | |||
NM_145273.3 | NP_660316.2 | 4 | 658 | |||
XM_006721721.2 | XP_006721784.1 | 4 | 1029 | |||
XM_006721722.2 | XP_006721785.1 | 3 | 927 | |||
XM_011524384.2 | XP_011522686.1 | 4 | 1029 | |||
GenBank mRNA | AK298112.1 | - | 3 | 558 | ||
AK309938.1 | - | 4 | 659 | |||
AY358364.1 | - | 4 | 613 | |||
BC025395.2 | - | 4 | 630 | 4827737 |