Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145285.2 | NP_660328.2 | 2 | 1959 | ||
GenBank mRNA | AK300856.1 | - | 2 | 1912 | ||
BC025788.1 | - | 3 | 1842 | 5229146 |