Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR9C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148897.2 | NP_683695.1 | 2, 3 | 712 | ||
GenBank mRNA | AK122782.1 | - | 2, 3 | 712 | ||
AY044434.1 | - | 2, 3 | 568 | |||
BC101551.1 | - | 2, 3 | 608 | 8069057 | ||
BC101553.1 | - | 2, 3 | 608 | 8069059 |