Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 7 | 1241 | ||
NM_001172767.2 | NP_001166238.1 | 7 | 955 | |||
NM_014491.3 | NP_055306.1 | 7 | 1244 | |||
NM_148898.3 | NP_683696.2 | 8 | 1319 | |||
NM_148899.3 | NP_683697.2 | 6 | 880 | |||
NM_148900.3 | NP_683698.2 | 8 | 1295 | |||
NR_033766.1 | 11 | 1457 | ||||
NR_033767.1 | 8 | 1302 | ||||
XM_017012801.1 | XP_016868290.1 | 10 | 1925 | |||
GenBank mRNA | AF337817.1 | - | 7 | 1020 | ||
AF467253.1 | - | 6 | 882 | |||
AF467255.1 | - | 1 | 97 | |||
AF467257.1 | - | 7 | 922 | |||
AF467258.1 | - | 6 | 867 | |||
AF467259.1 | - | 7 | 942 | |||
AK131266.1 | - | 11 | 1441 | |||
AK296957.1 | - | 8 | 1175 | |||
AY144615.1 | - | 8 | 1314 | |||
BC126104.1 | - | 8 | 976 | 8991820 | ||
BC143866.1 | - | 8 | 976 | 9052382 | ||
BC143867.1 | - | 7 | 915 | 9052383 | ||
CR749236.1 | - | 13 | 1470 | |||
DQ778626.1 | - | 4 | 594 | |||
KJ534839.1 | - | 7 | 945 | |||
KJ535079.1 | - | 6 | 1083 | |||
KJ535086.1 | - | 6 | 869 |