Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 9 | 1537 | ||
NM_001172767.2 | NP_001166238.1 | 9 | 1251 | |||
NM_014491.3 | NP_055306.1 | 9 | 1540 | |||
NM_148898.3 | NP_683696.2 | 10 | 1615 | |||
NM_148899.3 | NP_683697.2 | 8 | 1176 | |||
NM_148900.3 | NP_683698.2 | 10 | 1591 | |||
NR_033766.1 | 13 | 1753 | ||||
NR_033767.1 | 10 | 1598 | ||||
XM_017012801.1 | XP_016868290.1 | 12 | 2221 | |||
GenBank mRNA | AF337817.1 | - | 9 | 1316 | ||
AF467253.1 | - | 8 | 1178 | |||
AF467254.1 | - | 2 | 157 | |||
AF467255.1 | - | 3 | 393 | |||
AF467256.1 | - | 2 | 157 | |||
AF467258.1 | - | 8 | 1163 | |||
AF467259.1 | - | 9 | 1238 | |||
AK131266.1 | - | 13 | 1737 | |||
AK296957.1 | - | 10 | 1471 | |||
AY144615.1 | - | 10 | 1610 | |||
BC126104.1 | - | 10 | 1272 | 8991820 | ||
BC143866.1 | - | 10 | 1272 | 9052382 | ||
BC143867.1 | - | 9 | 1211 | 9052383 | ||
CR749236.1 | - | 15 | 1766 | |||
DQ778626.1 | - | 6 | 890 | |||
KJ534839.1 | - | 9 | 1241 | |||
KJ535079.1 | - | 8 | 1379 | |||
KJ535086.1 | - | 8 | 1165 |