Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 17 | 2658 | ||
NM_014491.3 | NP_055306.1 | 17 | 2661 | |||
NM_148898.3 | NP_683696.2 | 18 | 2736 | |||
NM_148900.3 | NP_683698.2 | 18 | 2712 | |||
NR_033766.1 | 20 | 2672 | ||||
NR_033767.1 | 18 | 2719 | ||||
XM_017012801.1 | XP_016868290.1 | 20 | 3342 | |||
GenBank mRNA | AF337817.1 | - | 17 | 2437 | ||
AK131266.1 | - | 20 | 2656 | |||
AK296957.1 | - | 18 | 2592 |