Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286139.1 | NP_001273068.1 | 2 | 288 | ||
NM_152313.3 | NP_689526.2 | 2 | 222 | |||
XM_017017174.1 | XP_016872663.1 | 2 | 226 | |||
XM_017017175.1 | XP_016872664.1 | 2 | 230 | |||
XM_017017176.1 | XP_016872665.1 | 2 | 211 | |||
XM_017017177.1 | XP_016872666.1 | 1 | 24 | |||
GenBank mRNA | AK096251.1 | - | 2 | 287 | ||
AY162216.1 | - | 2 | 176 | |||
BC047374.1 | - | 2 | 189 | 5313230 | ||
HY170772.1 | - | 2 | 222 |