Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286139.1 | NP_001273068.1 | 11 | 2430 | ||
NM_152313.3 | NP_689526.2 | 11 | 2393 | |||
XM_017017174.1 | XP_016872663.1 | 11 | 2384 | |||
XM_017017175.1 | XP_016872664.1 | 10 | 2310 | |||
XM_017017176.1 | XP_016872665.1 | 10 | 2262 | |||
XM_017017177.1 | XP_016872666.1 | 10 | 2153 | |||
GenBank mRNA | AJ295983.1 | - | 6 | 1783 | ||
AY162216.1 | - | 11 | 2347 |