Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172681.1 | NP_001166152.1 | 4 | 517 | ||
NM_152320.2 | NP_689533.2 | 5 | 690 | |||
XM_005268638.3 | XP_005268695.1 | 4 | 710 | |||
XM_005268640.4 | XP_005268697.1 | 4 | 690 | |||
XM_011537897.1 | XP_011536199.1 | 4 | 690 | |||
XM_011537898.1 | XP_011536200.1 | 4 | 618 | |||
XM_017018798.1 | XP_016874287.1 | 4 | 544 | |||
XM_017018799.1 | XP_016874288.1 | 3 | 424 | |||
GenBank mRNA | AK055857.1 | - | 4 | 593 | ||
AK092740.1 | - | 4 | 1117 | |||
AY842285.1 | - | 4 | 1144 | |||
BC018090.1 | - | 4 | 517 | 4791876 |