Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152329.3 | NP_689542.2 | 4 | 1440 | ||
NM_203467.1 | NP_982292.1 | 3 | 718 | |||
NR_037792.1 | 6 | 1660 | ||||
NR_037793.1 | 5 | 1548 | ||||
GenBank mRNA | AK293156.1 | - | 4 | 1159 | ||
AY052405.1 | - | 4 | 1126 | |||
BC030142.2 | - | 3 | 654 | |||
BC064133.1 | - | 5 | 1293 | |||
BC093697.1 | - | 3 | 420 | |||
BC112241.1 | - | 3 | 420 | |||
BC139921.1 | - | 4 | 1180 | 40148567 | ||
BQ059807.1 | - | 4 | 455 | |||
BX248298.1 | - | 4 | 1183 | |||
BX648029.1 | - | 6 | 1583 |