Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLHC1 SIRNA Assays ›
Gene Symbol:
CLHC1
Gene Name:
clathrin heavy chain linker domain containing 1
Gene Aliases:
C2orf63
Chromosome Location:
Chr.2: 55172547 - 55232563 on Build GRCh38
UniGene ID:
Hs.347014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110152
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLHC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135598.1
NP_001129070.1
3, 4
402
NM_152385.2
NP_689598.2
5, 6
853
XM_005264136.2
XP_005264193.1
4, 5
721
XM_006711937.3
XP_006712000.2
2, 3
160
XM_011532519.2
XP_011530821.1
5, 6
3746
XM_011532520.2
XP_011530822.1
5, 6
1024
XM_011532521.2
XP_011530823.1
6, 7
1180
XM_011532522.2
XP_011530824.1
6, 7
1167
XM_011532523.2
XP_011530825.1
5, 6
1256
XM_011532524.1
XP_011530826.1
4, 5
638
XM_011532525.2
XP_011530827.1
4, 5
704
XM_011532527.2
XP_011530829.1
5, 6
4348
XM_011532528.1
XP_011530830.1
4, 5
1248
XM_011532529.2
XP_011530831.1
3, 4
1207
XM_011532530.1
XP_011530832.1
3, 4
968
XM_017003333.1
XP_016858822.1
5, 6
3746
XM_017003334.1
XP_016858823.1
5, 6
1024
XM_017003335.1
XP_016858824.1
3, 4
3916
XM_017003336.1
XP_016858825.1
4, 5
1337
XM_017003337.1
XP_016858826.1
2, 3
426
XM_017003338.1
XP_016858827.1
2, 3
404
XM_017003339.1
XP_016858828.1
5, 6
3746
XM_017003343.1
XP_016858832.1
4, 5
1248
XM_017003344.1
XP_016858833.1
4, 5
1336
XM_017003345.1
XP_016858834.1
3, 4
1206
XM_017003346.1
XP_016858835.1
5, 6
3746
XM_017003348.1
XP_016858837.1
5, 6
3746
XM_017003349.1
XP_016858838.1
1, 2
3380
GenBank mRNA
AK315683.1
-
5, 6
811
BC029502.1
-
5, 6
853
5270720
BC036908.1
-
3, 4
402
5166064
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0