Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044385.2 | NP_001037850.1 | 13 | 1383 | ||
NM_152388.3 | NP_689601.2 | 13 | 1548 | |||
GenBank mRNA | AB053301.1 | - | 13 | 1550 | ||
AK090601.1 | - | 13 | 1241 | |||
AK125841.1 | - | 2 | 393 | |||
AK303954.1 | - | 13 | 1370 | |||
AK304395.1 | - | 11 | 1072 | |||
BC013730.1 | - | 5 | 466 | |||
BC029611.1 | - | 13 | 1296 | 5304046 | ||
BX538000.1 | - | 5 | 443 |