Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204298.1 | NP_001191227.1 | 5 | 1893 | ||
NM_152437.2 | NP_689650.1 | 5 | 1945 | |||
GenBank mRNA | AK023009.1 | - | 1 | 238 | ||
AK056034.1 | - | 5 | 1852 | |||
AL834266.1 | - | 6 | 2746 | |||
BC051696.1 | - | 5 | 1875 | 5276234 | ||
BX647762.1 | - | 6 | 1865 |