Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146154.1 | NP_001139626.1 | 5 | 610 | ||
NM_001146155.1 | NP_001139627.1 | 5 | 658 | |||
NM_152444.2 | NP_689657.1 | 5 | 700 | |||
GenBank mRNA | AK096410.1 | - | 5 | 650 | ||
AY346133.1 | - | 5 | 596 | |||
AY424308.1 | - | 5 | 596 | |||
BC059364.1 | - | 5 | 679 | 5301759 | ||
BX641118.1 | - | 5 | 516 | |||
DR158023.1 | - | 5 | 658 | |||
FJ224334.1 | - | 5 | 596 |