Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042474.2 | NP_001035939.1 | 5 | 603 | ||
NM_152477.4 | NP_689690.3 | 5 | 458 | |||
XM_011526512.2 | XP_011524814.1 | 5 | 671 | |||
XM_011526514.2 | XP_011524816.1 | 5 | 537 | |||
XM_017026341.1 | XP_016881830.1 | 6 | 1146 | |||
XM_017026342.1 | XP_016881831.1 | 4 | 366 | |||
XM_017026343.1 | XP_016881832.1 | 4 | 412 | |||
GenBank mRNA | AK057290.1 | - | 5 | 1190 | ||
AK094310.1 | - | 5 | 600 | |||
AK308212.1 | - | 4 | 376 | |||
BC068453.1 | - | 5 | 440 | 30343899 | ||
CB960081.1 | - | 5 | 467 | |||
DA473136.1 | - | 5 | 458 | |||
DB473200.1 | - | 5 | 451 |