Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278623.1 | NP_001265552.1 | 10 | 1371 | ||
NM_001278624.1 | NP_001265553.1 | 10 | 1509 | |||
NM_152995.5 | NP_694540.3 | 10 | 1411 | |||
NR_103795.1 | 10 | 1371 | ||||
GenBank mRNA | AB085695.1 | - | 10 | 1367 | ||
AB181916.1 | - | 10 | 1491 | |||
AK131285.1 | - | 8 | 957 | |||
AK302440.1 | - | 10 | 1369 | |||
BC051193.1 | - | 10 | 1342 | 5271401 |