Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143941.1 | NP_001137413.1 | 4 | 950 | ||
NM_001143942.1 | NP_001137414.1 | 4 | 966 | |||
NM_153020.2 | NP_694565.1 | 3 | 898 | |||
XM_005248932.3 | XP_005248989.1 | 5 | 1081 | |||
XM_011514387.2 | XP_011512689.1 | 4 | 867 | |||
XM_011514388.2 | XP_011512690.1 | 4 | 1192 | |||
XM_011514389.2 | XP_011512691.1 | 3 | 1141 | |||
GenBank mRNA | AK055391.1 | - | 3 | 897 | ||
AK095016.1 | - | 5 | 913 | |||
AK129865.1 | - | 5 | 884 | |||
AK293052.1 | - | 4 | 966 | |||
AK300062.1 | - | 2 | 695 | |||
AK300068.1 | - | 3 | 760 | |||
AK315856.1 | - | 4 | 966 | |||
AL832199.1 | - | 1 | 4946 | |||
AL833156.1 | - | 2 | 395 |