Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128165.1 | NP_001121637.1 | 7 | 1852 | ||
NM_153214.2 | NP_694946.2 | 8 | 1990 | |||
XM_006712260.3 | XP_006712323.1 | 8 | 2135 | |||
XM_011510585.2 | XP_011508887.1 | 9 | 1902 | |||
XM_011510587.2 | XP_011508889.1 | 10 | 1879 | |||
GenBank mRNA | AK094759.1 | - | 8 | 1990 | ||
AK124067.1 | - | 6 | 1388 | |||
BC035784.1 | - | 8 | 1943 | 5589120 | ||
BC038242.1 | - | 4 | 1185 |