Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153260.2 | NP_694992.2 | 6 | 872 | ||
GenBank mRNA | AK094131.1 | - | 6 | 866 | ||
AK094891.1 | - | 5 | 1106 | |||
AK309861.1 | - | 5 | 1437 | |||
BC058935.1 | - | 6 | 828 | 5578024 | ||
BX538324.1 | - | 5 | 1018 | |||
CN350663.1 | - | 3 | 322 | |||
CR749223.1 | - | 6 | 841 |