Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3MBTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173464.3 | NP_775735.2 | 10, 11 | 993 | ||
XM_005258166.1 | XP_005258223.1 | 10, 11 | 952 | |||
XM_006722364.2 | XP_006722427.1 | 10, 11 | 952 | |||
XM_011525757.1 | XP_011524059.1 | 10, 11 | 952 | |||
XM_011525758.1 | XP_011524060.1 | 10, 11 | 966 | |||
XM_011525759.1 | XP_011524061.1 | 10, 11 | 973 | |||
XM_011525760.1 | XP_011524062.1 | 10, 11 | 952 | |||
XM_011525761.1 | XP_011524063.1 | 10, 11 | 966 | |||
XM_011525762.1 | XP_011524064.1 | 10, 11 | 952 | |||
XM_011525763.2 | XP_011524065.1 | 10, 11 | 952 | |||
XM_011525765.2 | XP_011524067.1 | 10, 11 | 952 | |||
XM_017026074.1 | XP_016881563.1 | 10, 11 | 952 | |||
XM_017026075.1 | XP_016881564.1 | 10, 11 | 952 | |||
XM_017026076.1 | XP_016881565.1 | 10, 11 | 973 | |||
XM_017026077.1 | XP_016881566.1 | 10, 11 | 952 | |||
GenBank mRNA | AK091075.1 | - | 3, 4 | 282 | ||
AK093255.1 | - | 10, 11 | 952 | |||
BC039316.1 | - | 10, 11 | 985 | 5298402 |