Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173484.3 | NP_775755.3 | 4 | 2201 | ||
XM_005270465.3 | XP_005270522.2 | 5 | 1825 | |||
XM_011540700.1 | XP_011539002.1 | 7 | 2897 | |||
GenBank mRNA | BC049844.1 | - | 4 | 2201 | 5166200 |