Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199148.1 | NP_001186077.1 | 11 | 1340 | ||
NM_173512.2 | NP_775783.1 | 10 | 1274 | |||
XM_005246350.4 | XP_005246407.1 | 12 | 1493 | |||
XM_006712337.3 | XP_006712400.1 | 11 | 1489 | |||
XM_011510737.2 | XP_011509039.1 | 11 | 1585 | |||
XM_011510742.2 | XP_011509044.1 | 9 | 1281 | |||
XM_017003455.1 | XP_016858944.1 | 12 | 1729 | |||
XM_017003456.1 | XP_016858945.1 | 11 | 1663 | |||
XM_017003457.1 | XP_016858946.1 | 11 | 1654 | |||
XM_017003458.1 | XP_016858947.1 | 11 | 1614 | |||
XM_017003459.1 | XP_016858948.1 | 10 | 1352 | |||
XM_017003463.1 | XP_016858952.1 | 11 | 1126 | |||
XM_017003464.1 | XP_016858953.1 | 8 | 1338 | |||
XM_017003465.1 | XP_016858954.1 | 7 | 866 | |||
GenBank mRNA | AK097141.1 | - | 10 | 1274 | ||
AK293998.1 | - | 11 | 1301 | |||
BC125157.1 | - | 13 | 1267 | 40120878 |