Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002090.2 | NP_002081.2 | 4 | 643 | ||
GenBank mRNA | AK311367.1 | - | 2 | 1606 | ||
BC016308.1 | - | 4 | 563 | 4070983 | ||
BC065743.1 | - | 4 | 563 | 4717346 | ||
M36821.1 | - | 4 | 556 | |||
X53800.1 | - | 4 | 515 |