Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173658.2 | NP_775929.2 | 3 | 1896 | ||
XM_005265090.4 | XP_005265147.1 | 2 | 1994 | |||
XM_006713117.3 | XP_006713180.1 | 3 | 1995 | |||
XM_011533622.2 | XP_011531924.1 | 3 | 2199 | |||
GenBank mRNA | AK094189.1 | - | 3 | 1896 | ||
BX538171.1 | - | 1 | 1321 |