Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136230.2 | NP_001129702.1 | 2 | 391 | ||
NM_178135.4 | NP_835236.2 | 3 | 499 | |||
GenBank mRNA | AB073347.1 | - | 3 | 468 | ||
AK292784.1 | - | 3 | 473 | |||
AY186249.1 | - | 3 | 479 | |||
AY186250.1 | - | 2 | 371 | |||
AY268355.1 | - | 3 | 449 | |||
AY358575.1 | - | 3 | 499 | |||
BC112303.1 | - | 3 | 458 | 8327771 | ||
BC112305.1 | - | 3 | 458 | 8327773 | ||
DR004209.1 | - | 3 | 504 |