Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110426
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204148.2
NP_001191077.1
6
900
NM_001321869.1
NP_001308798.1
8
1182
NM_001321870.1
NP_001308799.1
8
1173
NM_001321874.1
NP_001308803.1
7
1103
NM_001321875.1
NP_001308804.1
7
1109
NM_001321876.1
NP_001308805.1
7
1097
NM_001321878.1
NP_001308807.1
7
1094
NM_001321884.1
NP_001308813.1
7
1097
NM_001321886.1
NP_001308815.1
8
1153
NM_001321887.1
NP_001308816.1
8
987
NM_001321890.1
NP_001308819.1
7
1088
NM_001321896.1
NP_001308825.1
8
1272
NM_001321899.1
NP_001308828.1
8
1000
NM_001321900.1
NP_001308829.1
7
1094
NM_001321901.1
NP_001308830.1
7
928
NM_001321902.1
NP_001308831.1
7
934
NM_014707.3
NP_055522.1
7
925
NM_058176.2
NP_478056.1
6
780
NM_178423.2
NP_848510.1
7
925
NM_178425.3
NP_848512.1
6
789
XM_011515629.2
XP_011513931.1
7
883
XM_011515631.2
XP_011513933.1
8
1197
XM_011515635.2
XP_011513937.1
8
1749
XM_011515636.2
XP_011513938.1
7
1470
XM_011515637.2
XP_011513939.1
7
894
XM_011515638.2
XP_011513940.1
7
1464
XM_011515639.2
XP_011513941.1
7
1661
XM_011515641.2
XP_011513943.1
8
1746
XM_011515642.2
XP_011513944.1
6
2475
XM_011515644.2
XP_011513946.1
8
1721
XM_011515645.2
XP_011513947.1
6
2460
XM_011515646.2
XP_011513948.1
6
2451
XM_011515648.2
XP_011513950.1
7
1479
XM_011515649.2
XP_011513951.1
7
1683
XM_017012826.1
XP_016868315.1
7
1455
XM_017012827.1
XP_016868316.1
8
1538
XM_017012829.1
XP_016868318.1
7
858
XM_017012830.1
XP_016868319.1
7
849
XM_017012832.1
XP_016868321.1
7
858
GenBank mRNA
AB018287.1
-
7
889
AF124924.1
-
4
447
AJ459808.1
-
7
889
AK304343.1
-
6
820
AY032737.1
-
6
739
AY032738.1
-
6
739
AY197371.1
-
6
748
BC150328.1
-
7
900
8860495
BC152405.1
-
7
889
8862508
CR749550.1
-
6
856
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline