Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163941.1 | NP_001157413.1 | 15 | 1962 | ||
NM_178559.5 | NP_848654.3 | 6 | 936 | |||
XM_011515367.2 | XP_011513669.1 | 7 | 882 | |||
GenBank mRNA | AB353947.1 | - | 15 | 1962 | ||
AF399931.1 | - | 4 | 450 | |||
AY090613.1 | - | 6 | 857 | |||
AY230001.1 | - | 15 | 1855 | |||
AY234788.1 | - | 6 | 857 | |||
AY851365.1 | - | 6 | 743 | |||
BC104894.1 | - | 6 | 686 | 8143897 | ||
BC104920.1 | - | 6 | 684 | 8143923 | ||
GU437216.1 | - | 3 | 390 | |||
GU437217.1 | - | 3 | 390 |