Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163941.1 | NP_001157413.1 | 25 | 3282 | ||
NM_178559.5 | NP_848654.3 | 16 | 2256 | |||
XM_011515367.2 | XP_011513669.1 | 17 | 2202 | |||
GenBank mRNA | AB353947.1 | - | 25 | 3282 | ||
AF319622.1 | - | 11 | 1322 | |||
AF399931.1 | - | 14 | 1770 | |||
AK127141.1 | - | 5 | 546 | |||
AY090613.1 | - | 16 | 2177 | |||
AY230001.1 | - | 25 | 3175 | |||
AY234788.1 | - | 16 | 2177 | |||
AY851365.1 | - | 16 | 2063 | |||
BC104894.1 | - | 16 | 2006 | 8143897 | ||
BC104920.1 | - | 16 | 2004 | 8143923 | ||
GU437216.1 | - | 13 | 1710 | |||
GU437217.1 | - | 13 | 1710 |