Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180989.5 | NP_851320.1 | 2 | 317 | ||
GenBank mRNA | AK126794.1 | - | 2 | 222 | ||
AK289850.1 | - | 2 | 564 | |||
BC033906.1 | - | 2 | 292 | |||
BC052243.1 | - | 2 | 264 | 6083073 | ||
CD110068.1 | - | 2 | 344 |