Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180989.5 | NP_851320.1 | 9 | 2166 | ||
GenBank mRNA | AK024059.1 | - | 1 | 594 | ||
AK126794.1 | - | 9 | 2070 | |||
BC033906.1 | - | 8 | 1960 |