Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258041.2 | NP_001244970.1 | 3 | 656 | ||
NM_001289092.1 | NP_001276021.1 | 3 | 656 | |||
NM_001289094.1 | NP_001276023.1 | 3 | 569 | |||
NM_002109.5 | NP_002100.2 | 3 | 656 | |||
GenBank mRNA | AK000498.1 | - | 3 | 336 | ||
AK124831.1 | - | 2 | 1846 | |||
AK225776.1 | - | 3 | 306 | |||
AK293154.1 | - | 3 | 270 | |||
AK293531.1 | - | 318 | ||||
AK307669.1 | - | 3 | 249 | |||
AY995220.1 | - | 3 | 233 | |||
BC011807.2 | - | 3 | 263 | 4331277 | ||
BC080514.1 | - | 3 | 261 | 2821065 | ||
DC394573.1 | - | 3 | 656 | |||
X05345.1 | - | 3 | 310 | |||
Z11518.1 | - | 3 | 791 |