Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181776.2 | NP_861441.2 | 10 | 3352 | ||
GenBank mRNA | AY162214.1 | - | 10 | 3320 |