Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HKR1 SIRNA Assays ›
Gene Symbol:
HKR1
Gene Name:
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases:
ZNF875
Chromosome Location:
Chr.19: 37317900 - 37364455 on Build GRCh38
UniGene ID:
Hs.657027
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110481
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HKR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181786.2
NP_861451.1
6
1286
XM_017026672.1
XP_016882161.1
4
1474
XM_017026673.1
XP_016882162.1
4
1540
XM_017026674.1
XP_016882163.1
8
1840
XM_017026675.1
XP_016882164.1
8
1837
XM_017026676.1
XP_016882165.1
5
1135
XM_017026677.1
XP_016882166.1
5
1122
XM_017026678.1
XP_016882167.1
5
1132
XM_017026679.1
XP_016882168.1
5
1119
XM_017026680.1
XP_016882169.1
8
1502
XM_017026681.1
XP_016882170.1
7
1450
XM_017026682.1
XP_016882171.1
8
1499
XM_017026683.1
XP_016882172.1
7
1361
XM_017026684.1
XP_016882173.1
6
1223
XM_017026685.1
XP_016882174.1
4
1068
XM_017026686.1
XP_016882175.1
7
1358
XM_017026687.1
XP_016882176.1
6
1220
XM_017026688.1
XP_016882177.1
4
3292
XM_017026689.1
XP_016882178.1
6
1306
XM_017026690.1
XP_016882179.1
4
1086
XM_017026691.1
XP_016882180.1
3
866
XM_017026692.1
XP_016882181.1
8
1436
XM_017026693.1
XP_016882182.1
9
1557
XM_017026694.1
XP_016882183.1
9
1507
XM_017026695.1
XP_016882184.1
8
1479
XM_017026696.1
XP_016882185.1
7
1331
XM_017026697.1
XP_016882186.1
6
1253
XM_017026698.1
XP_016882187.1
6
1231
XM_017026699.1
XP_016882188.1
4
4067
XM_017026700.1
XP_016882189.1
6
1232
XM_017026701.1
XP_016882190.1
8
1430
XM_017026702.1
XP_016882191.1
7
1352
XM_017026703.1
XP_016882192.1
5
1129
XM_017026704.1
XP_016882193.1
5
1191
XM_017026705.1
XP_016882194.1
2
1188
XM_017026707.1
XP_016882196.1
5
1604
XM_017026708.1
XP_016882197.1
5
1607
GenBank mRNA
AB013897.1
-
5
1133
AK056243.1
-
6
1220
AK128116.1
-
9
1839
AK223616.1
-
5
1126
AK299969.1
-
5
1100
AK307852.1
-
1
844
BC004513.2
-
1
308
BC017256.2
-
6
1286
3356680
BC053845.1
-
4
1123
5493685
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline