Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286668.1 | NP_001273597.1 | 14 | 2981 | ||
NM_001286669.1 | NP_001273598.1 | 14 | 3132 | |||
NM_001286670.1 | NP_001273599.1 | 12 | 2656 | |||
NM_001286671.1 | NP_001273600.1 | 12 | 2834 | |||
NM_001323890.1 | NP_001310819.1 | 16 | 3370 | |||
NM_001323892.1 | NP_001310821.1 | 14 | 3089 | |||
NM_001323893.1 | NP_001310822.1 | 13 | 2968 | |||
NM_001323899.1 | NP_001310828.1 | 14 | 3146 | |||
NM_181806.3 | NP_861522.2 | 15 | 3253 | |||
XM_017007740.1 | XP_016863229.1 | 14 | 3082 | |||
XM_017007741.1 | XP_016863230.1 | 16 | 3329 | |||
XM_017007742.1 | XP_016863231.1 | 13 | 3029 | |||
XM_017007743.1 | XP_016863232.1 | 13 | 2918 | |||
XM_017007744.1 | XP_016863233.1 | 12 | 2836 | |||
GenBank mRNA | AF516672.1 | - | 14 | 2896 | ||
AK294332.1 | - | 12 | 2618 | |||
AK303733.1 | - | 13 | 2921 | |||
AK304309.1 | - | 15 | 3244 | |||
AK316296.1 | - | 15 | 3234 | |||
AY314787.1 | - | 15 | 3216 | |||
AY422212.1 | - | 15 | 3168 | |||
BC071815.1 | - | 1 | 167 | |||
BC142709.1 | - | 12 | 2770 | 40148820 | ||
BX640635.1 | - | 10 | 4084 |