Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMIGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143668.1 | NP_001137140.1 | 3 | 2050 | ||
NM_181847.4 | NP_862830.1 | 2 | 1948 | |||
XM_005268837.3 | XP_005268894.1 | 3 | 2930 | |||
GenBank mRNA | AB079074.1 | - | 1 | 679 | ||
AB080610.1 | - | 1 | 1458 | |||
AK091761.1 | - | 2 | 1771 | |||
AK093111.1 | - | 1 | 888 | |||
AL833007.1 | - | 3 | 1746 | |||
AY237005.1 | - | 1 | 1395 | |||
AY454159.1 | - | 2 | 1863 | |||
BC014103.2 | - | 1 | 662 | |||
BC047595.1 | - | 2 | 1863 | 4828334 | ||
BC095477.1 | - | 3 | 2050 | 30387316 |