Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318246.1 | NP_001305175.1 | 8 | 1014 | ||
NM_194071.3 | NP_919047.2 | 8 | 1427 | |||
XM_005250541.4 | XP_005250598.1 | 8 | 1806 | |||
XM_017012540.1 | XP_016868029.1 | 9 | 2829 | |||
XM_017012541.1 | XP_016868030.1 | 9 | 1392 | |||
GenBank mRNA | AJ549092.1 | - | 8 | 1380 | ||
AJ549387.1 | - | 8 | 1380 | |||
AK131517.1 | - | 8 | 1393 | |||
AK293048.1 | - | 8 | 1359 | |||
BC110813.1 | - | 8 | 1363 | 3923538 | ||
BX649143.1 | - | 8 | 1019 | |||
CR749563.1 | - | 7 | 1107 |