Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015881.1 | NP_001015881.1 | 3 | 1533 | ||
NM_001318468.1 | NP_001305397.1 | 4 | 2001 | |||
NM_001318470.1 | NP_001305399.1 | 4 | 1987 | |||
NM_004089.3 | NP_004080.2 | 3 | 1738 | |||
NM_198057.2 | NP_932174.1 | 3 | 2054 | |||
XM_005262099.1 | XP_005262156.1 | 5 | 2015 | |||
XM_005262100.1 | XP_005262157.1 | 4 | 1890 | |||
XM_005262102.1 | XP_005262159.1 | 4 | 1883 | |||
XM_005262103.3 | XP_005262160.1 | 4 | 1993 | |||
XM_011530884.1 | XP_011529186.1 | 4 | 2089 | |||
XM_017029335.1 | XP_016884824.1 | 4 | 1825 | |||
GenBank mRNA | AB025432.1 | - | 3 | 1722 | ||
AF153603.1 | - | 3 | 1715 | |||
AF183393.1 | - | 3 | 1724 | |||
AF228339.1 | - | 3 | 1728 | |||
AK092645.1 | - | 4 | 1940 | |||
AK092669.1 | - | 3 | 2026 | |||
AK127938.1 | - | 3 | 1678 | |||
AL110191.1 | - | 3 | 1736 | |||
AY007119.1 | - | 3 | 1637 | |||
BC018148.1 | - | 3 | 1420 | |||
BC072446.1 | - | 3 | 1732 | 6165765 | ||
BX647854.1 | - | 4 | 1987 | |||
CR933650.1 | - | 3 | 1740 |