Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031679.2 | NP_001026849.1 | 3 | 450 | ||
NM_001193460.1 | NP_001180389.1 | 4 | 614 | |||
NM_001193461.1 | NP_001180390.1 | 3 | 312 | |||
NM_198080.3 | NP_932346.1 | 2 | 323 | |||
GenBank mRNA | AK293084.1 | - | 3 | 420 | ||
AK299065.1 | - | 2 | 262 | |||
AK316365.1 | - | 2 | 294 | |||
AL833622.1 | - | 3 | 368 | |||
AY358229.1 | - | 2 | 258 | |||
BC040053.1 | - | 4 | 515 | 6025598 | ||
BX500552.1 | - | 3 | 314 | |||
BX648776.1 | - | 3 | 369 | |||
DC357690.1 | - | 2 | 323 |